Learn More
Medchemexpress LLC Boc-NH-C4-acid | 27219-07-4 | MFCD00076903 | 99.9% | 217.26 g/mol | C10H19NO4 | 1 G
SDP
Catalog No. 5000304051
Encompass_Preferred
Boc-NH-C4-acid is a Boc-protected 5-aminopentanoic acid used as an alkyl/ether PROTAC linker and building block in medicinal chemistry. Supplied as a solid with high reported purity, it functions as a protected amino acid intermediate for linker construction, peptide coupling, and conjugation workflows in research applications.
  • High reported purity (approximately 99.85%).
  • Provides Boc protection for the amino group, enabling selective deprotection.
  • Contains a carboxylic acid handle for standard coupling chemistries.
  • Suitable for synthesis of PROTAC linkers and other conjugates.
  • Available in multiple laboratory sizes to support scale-up.
  • Intended for research use only.
Catalog No. 50-003-04051 Supplier Medchemexpress LLC Supplier No. HYW0140991G
Boc-NH-C4-acid is a Boc-protected 5-aminopentanoic acid used as an alkyl/ether PROTAC linker and building block in medicinal chemistry. Supplied as a solid with high reported purity, it functions as a protected amino acid intermediate for linker construction, peptide coupling, and conjugation workflows in research applications.
  • High reported purity (approximately 99.85%).
  • Provides Boc protection for the amino group, enabling selective deprotection.
  • Contains a carboxylic acid handle for standard coupling chemistries.
  • Suitable for synthesis of PROTAC linkers and other conjugates.
  • Available in multiple laboratory sizes to support scale-up.
  • Intended for research use only.