Learn More
Medchemexpress LLC Boc-NH-C4-Br | 164365-88-2 | 252.15 | 25 G
SDP
Catalog No. 5000449986
Encompass_Preferred
Boc-NH-C4-Br is an alkyl chain-based PROTAC linker primarily used in the synthesis of PROTACs (Proteolysis Targeting Chimeras). PROTACs are designed with two distinct ligands connected by a linker. One ligand is specific for an E3 ubiquitin ligase, and the other targets a protein of interest.
  • Leverages the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins
Catalog No. 50-004-49986 Supplier Medchemexpress LLC Supplier No. 25GHYW007803
Boc-NH-C4-Br is an alkyl chain-based PROTAC linker primarily used in the synthesis of PROTACs (Proteolysis Targeting Chimeras). PROTACs are designed with two distinct ligands connected by a linker. One ligand is specific for an E3 ubiquitin ligase, and the other targets a protein of interest.
  • Leverages the intracellular ubiquitin-proteasome system
  • Enables selective degradation of target proteins