Learn More
Medchemexpress LLC Boc-NH-C4-Br | 164365-88-2 | 252.15 | 5 G
SDP
Catalog No. 5000446735
Encompass_Preferred
Boc-NH-C4-Br (MedChemExpress HY-W007803) is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTACs. This product is intended for research use only.
  • Alkyl chain-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Solid below 30°C, liquid above 32°C
  • White to off-white appearance
  • Soluble in DMSO at 100 mg/mL
  • Requires ultrasonic and warming to 60°C for dissolution in DMSO
  • Solutions should be aliquoted to prevent inactivation from repeated freeze-thaw cycles
Catalog No. 50-004-46735 Supplier Medchemexpress LLC Supplier No. HYW0078035G
Boc-NH-C4-Br (MedChemExpress HY-W007803) is an alkyl chain-based PROTAC linker utilized in the synthesis of PROTACs. This product is intended for research use only.
  • Alkyl chain-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Solid below 30°C, liquid above 32°C
  • White to off-white appearance
  • Soluble in DMSO at 100 mg/mL
  • Requires ultrasonic and warming to 60°C for dissolution in DMSO
  • Solutions should be aliquoted to prevent inactivation from repeated freeze-thaw cycles