Learn More
Medchemexpress LLC Boc-NH-ethyl-SS-propionic acid | 485800-27-9 | CB73363888 | 95.0% | 281.39 | C10H19NO4S2 | 100 MG
SDP
Catalog No. 5000204031
Encompass_Preferred
Boc-NH-ethyl-SS-propionic acid is a cleavable disulfide-containing linker used to prepare antibody-drug conjugates. The molecule features a Boc-protected amine and an ethyl-disulfide-propionic acid spacer that enables reductive release of attached payloads. Supplied as a white to off-white solid at 95.0% purity, it is intended for research use only.
  • Cleavable disulfide linker for reductive payload release.
  • Boc-protected amine suitable for amide or carbamate formation.
  • White to off-white solid with specified purity of 95.0%.
  • Storage: keep at 4°C under nitrogen; in solvent, -80°C for 6 months, -20°C for 1 month.
  • Intended for research use only.
Catalog No. 50-002-04031 Supplier Medchemexpress LLC Supplier No. HY140116100MG
Boc-NH-ethyl-SS-propionic acid is a cleavable disulfide-containing linker used to prepare antibody-drug conjugates. The molecule features a Boc-protected amine and an ethyl-disulfide-propionic acid spacer that enables reductive release of attached payloads. Supplied as a white to off-white solid at 95.0% purity, it is intended for research use only.
  • Cleavable disulfide linker for reductive payload release.
  • Boc-protected amine suitable for amide or carbamate formation.
  • White to off-white solid with specified purity of 95.0%.
  • Storage: keep at 4°C under nitrogen; in solvent, -80°C for 6 months, -20°C for 1 month.
  • Intended for research use only.