Learn More
Medchemexpress LLC Boc-NH-ethyl-SS-propionic acid | 485800-27-9 | MFCD19443373 | 95.0% | 281.39 g/mol | C10H19NO4S2 | 250 MG
SDP
Catalog No. 5000203666
Encompass_Preferred
Boc-NH-ethyl-SS-propionic acid is a cleavable disulfide-containing linker used for conjugating small-molecule payloads to antibodies during antibody-drug conjugate (ADC) synthesis. It features a Boc-protected aminoethyl spacer and a terminal carboxylic acid that enable coupling reactions and a reducible disulfide bond for intracellular release.
  • Cleavable disulfide linker for intracellular payload release.
  • Provides a Boc-protected aminoethyl spacer for controlled conjugation.
  • Terminal carboxylic acid enables amide coupling to payloads or carriers.
  • High purity suitable for research and medicinal chemistry workflows.
  • Solid form, soluble in common organic solvents such as DMSO and DMF.
  • Stable when stored at 4°C under inert atmosphere; protect from moisture and light.
Catalog No. 50-002-03666 Supplier Medchemexpress LLC Supplier No. HY140116250MG
Boc-NH-ethyl-SS-propionic acid is a cleavable disulfide-containing linker used for conjugating small-molecule payloads to antibodies during antibody-drug conjugate (ADC) synthesis. It features a Boc-protected aminoethyl spacer and a terminal carboxylic acid that enable coupling reactions and a reducible disulfide bond for intracellular release.
  • Cleavable disulfide linker for intracellular payload release.
  • Provides a Boc-protected aminoethyl spacer for controlled conjugation.
  • Terminal carboxylic acid enables amide coupling to payloads or carriers.
  • High purity suitable for research and medicinal chemistry workflows.
  • Solid form, soluble in common organic solvents such as DMSO and DMF.
  • Stable when stored at 4°C under inert atmosphere; protect from moisture and light.