Learn More
Medchemexpress LLC Boc-NH-PEG-amine | ≥98.0% | 100 MG
SDP
Catalog No. 5000356475
Encompass_Preferred
Boc-NH-PEG-amine (MW 5000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that link an E3 ubiquitin ligase ligand to a target protein ligand, utilizing the intracellular ubiquitin-proteasome system for selective degradation of target proteins. This product is for research use only.
  • PEG-based linker for PROTAC synthesis.
  • Enables the degradation of target proteins.
  • Utilizes the intracellular ubiquitin-proteasome system.
  • Contains two ligands: E3 ubiquitin ligase and target protein.
Catalog No. 50-003-56475 Supplier Medchemexpress LLC Supplier No. HY140727100MG
Boc-NH-PEG-amine (MW 5000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs are compounds that link an E3 ubiquitin ligase ligand to a target protein ligand, utilizing the intracellular ubiquitin-proteasome system for selective degradation of target proteins. This product is for research use only.
  • PEG-based linker for PROTAC synthesis.
  • Enables the degradation of target proteins.
  • Utilizes the intracellular ubiquitin-proteasome system.
  • Contains two ligands: E3 ubiquitin ligase and target protein.