Learn More
Medchemexpress LLC Boc-NH-PEG-amine (MW 5000) | ≥98.0% | 50 MG
SDP
Catalog No. 5000353131
Encompass_Preferred
Boc-NH-PEG-amine (MW 5000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs utilize an intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein. This product is provided as a solid, white to off-white appearance, with a purity of ≥98.0%. It is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system for selective protein degradation
  • Solid, white to off-white appearance
  • Purity of ≥98.0%
  • Average molecular weight of 5000
  • Stable under recommended storage conditions
  • Available in various quantities for research
Catalog No. 50-003-53131 Supplier Medchemexpress LLC Supplier No. HY14072750MG
Boc-NH-PEG-amine (MW 5000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs utilize an intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase and a ligand for the target protein. This product is provided as a solid, white to off-white appearance, with a purity of ≥98.0%. It is intended for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system for selective protein degradation
  • Solid, white to off-white appearance
  • Purity of ≥98.0%
  • Average molecular weight of 5000
  • Stable under recommended storage conditions
  • Available in various quantities for research