Learn More
Medchemexpress LLC Boc-NH-PEG1-OH | 26690-80-2 | 161.20 | 1 G
SDP
Catalog No. 5000460369
Encompass_Preferred
Boc-NH-PEG1-OH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein, and they exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
Catalog No. 50-004-60369 Supplier Medchemexpress LLC Supplier No. HYW0073221G
Boc-NH-PEG1-OH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein, and they exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only