Learn More
Medchemexpress LLC Boc-NH-PEG1-OH | 26690-80-2 | 161.20 | 25 G
SDP
Catalog No. 5000460286
Encompass_Preferred
Boc-NH-PEG1-OH is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It is intended for research purposes only and not sold to patients. This linker facilitates the creation of PROTACs, which are molecules that recruit an E3 ubiquitin ligase to a target protein, leading to the target protein's ubiquitination and subsequent degradation by the proteasome.
  • Used in PROTAC synthesis
  • Facilitates degradation of target proteins
  • Utilizes intracellular ubiquitin-proteasome system
Catalog No. 50-004-60286 Supplier Medchemexpress LLC Supplier No. HYW00732210G
Boc-NH-PEG1-OH is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It is intended for research purposes only and not sold to patients. This linker facilitates the creation of PROTACs, which are molecules that recruit an E3 ubiquitin ligase to a target protein, leading to the target protein's ubiquitination and subsequent degradation by the proteasome.
  • Used in PROTAC synthesis
  • Facilitates degradation of target proteins
  • Utilizes intracellular ubiquitin-proteasome system