Learn More
Medchemexpress LLC Boc-NH-PEG1-OH | 26690-80-2 | 161.20 | 5 G
SDP
Catalog No. 5000460370
Encompass_Preferred
Boc-NH-PEG1-OH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-004-60370 Supplier Medchemexpress LLC Supplier No. HYW0073225G
Boc-NH-PEG1-OH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins