Learn More
Medchemexpress LLC HY-W007322 500mg Medchemexpress, Boc-NH-PEG1-OH CAS:26690-80-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW007322500MG
Encompass_Preferred
Medchemexpress, HY-W007322 500mg Boc-NH-PEG1-OH CAS:26690-80-2 Boc-NH-PEG1-OH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3235