Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-NH-PEG12-CH2CH2COOH | 1415981-79-1 | 98.0% | C32H63NO16 | 100 MG
SDP

Catalog No. 5000353489
Encompass_Preferred

Boc-NH-PEG12-CH2CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It is intended for research purposes only.

  • Used in the synthesis of PROTACs
  • 98.0% purity
  • Molecular weight of 717.84
  • Appears as a colorless to light yellow liquid
  • Store at -20°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-003-53489 Supplier Medchemexpress LLC Supplier No. HY140472100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-NH-PEG12-CH2CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It is intended for research purposes only.

  • Used in the synthesis of PROTACs
  • 98.0% purity
  • Molecular weight of 717.84
  • Appears as a colorless to light yellow liquid
  • Store at -20°C, protected from light
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.