Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-NH-PEG12-CH2CH2COOH | 1415981-79-1 | 98.0% | C32H63NO16 | 50 MG
SDP

Catalog No. 5000357035
Encompass_Preferred

Boc-NH-PEG12-CH2CH2COOH is a PEG-based PROTAC linker primarily utilized in the synthesis of PROTACs. These innovative molecules leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by employing two different ligands connected by a linker: one binds to an E3 ubiquitin ligase, and the other targets the protein intended for degradation.

  • Utilized in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Supplied as a colorless to light yellow liquid
  • Density of 1.121 g/cm³
  • Recommended storage at -20°C, protected from light
  • For longer periods in solvent, store at -80°C
  • Exhibits good solubility in DMSO (100 mg/mL)
  • Intended for research use only

Catalog No. 50-003-57035 Supplier Medchemexpress LLC Supplier No. HY14047250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-NH-PEG12-CH2CH2COOH is a PEG-based PROTAC linker primarily utilized in the synthesis of PROTACs. These innovative molecules leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by employing two different ligands connected by a linker: one binds to an E3 ubiquitin ligase, and the other targets the protein intended for degradation.

  • Utilized in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • Supplied as a colorless to light yellow liquid
  • Density of 1.121 g/cm³
  • Recommended storage at -20°C, protected from light
  • For longer periods in solvent, store at -80°C
  • Exhibits good solubility in DMSO (100 mg/mL)
  • Intended for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.