Learn More
Medchemexpress LLC Boc-NH-PEG2 | 139115-91-6 | 205.25 | 1 G
SDP
Catalog No. 5000449679
Encompass_Preferred
This PEG-based PROTAC linker can be used in the synthesis of PROTACs. PROTACs are compounds containing two different ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the desired protein. These compounds utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs degrade target proteins using the ubiquitin-proteasome system
  • PROTACs consist of two different ligands connected by a linker
Catalog No. 50-004-49679 Supplier Medchemexpress LLC Supplier No. HYW0048961G
This PEG-based PROTAC linker can be used in the synthesis of PROTACs. PROTACs are compounds containing two different ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets the desired protein. These compounds utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs degrade target proteins using the ubiquitin-proteasome system
  • PROTACs consist of two different ligands connected by a linker