Learn More
Medchemexpress LLC Boc-NH-PEG2-C2-NH2 | 153086-78-3 | 248.32 | 500 MG
SDP
Catalog No. 5000445572
Encompass_Preferred
Boc-NH-PEG2-C2-NH2, also known as PROTAC Linker 13, is a PEG-based PROTAC linker essential for the synthesis of PROTACs. These molecules are designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Utilized in the synthesis of PROTACs
  • Peg-based linker
  • Supports targeted protein degradation research
Catalog No. 50-004-45572 Supplier Medchemexpress LLC Supplier No. HYW008474500MG
Boc-NH-PEG2-C2-NH2, also known as PROTAC Linker 13, is a PEG-based PROTAC linker essential for the synthesis of PROTACs. These molecules are designed to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Utilized in the synthesis of PROTACs
  • Peg-based linker
  • Supports targeted protein degradation research