Learn More
Medchemexpress LLC Boc-NH-PEG2-C2-NHS ester | 2183440-73-3 | ≥98.0% | C16H26N2O8 | 500 MG
SDP
Catalog No. 5000354590
Encompass_Preferred
Boc-NH-PEG2-C2-NHS ester is an alkyl/ether-based PROTAC linker used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins, functioning by connecting two ligands: one for an E3 ubiquitin ligase and one for the target protein. This compound appears as a white to off-white solid.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables selective degradation of target proteins
  • Appears as a white to off-white solid
  • For research use only
Catalog No. 50-003-54590 Supplier Medchemexpress LLC Supplier No. HY141117500MG
Boc-NH-PEG2-C2-NHS ester is an alkyl/ether-based PROTAC linker used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). PROTACs are compounds designed to exploit the intracellular ubiquitin-proteasome system for selective degradation of target proteins, functioning by connecting two ligands: one for an E3 ubiquitin ligase and one for the target protein. This compound appears as a white to off-white solid.
  • Alkyl/ether-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Enables selective degradation of target proteins
  • Appears as a white to off-white solid
  • For research use only