Learn More
Medchemexpress LLC Boc-NH-PEG2-CH2CH2COOH | 1365655-91-9 | MFCD22056304 | 98.0% | 277.31 g·mol-1 | C12H23NO6 | 500 MG
SDP
Catalog No. 5000300124
Encompass_Preferred
Boc-NH-PEG2-CH2CH2COOH is a Boc-protected, PEG-based linker for assembling PROTAC molecules. It combines a two-unit ethylene glycol spacer with a Boc-protected amino terminus and a terminal propionic acid, enabling versatile conjugation chemistry for connecting target ligands and E3 ligase ligands in small-molecule degraders.
  • Boc-protected amino terminus for selective deprotection and coupling.
  • Two-unit PEG spacer provides flexibility and appropriate linker length.
  • Terminal carboxylic acid enables standard amide or ester bond formation.
  • High purity suitable for research-grade synthesis workflows.
  • Good solubility in DMSO for easy stock solution preparation.
Catalog No. 50-003-00124 Supplier Medchemexpress LLC Supplier No. HYW025812500MG
Boc-NH-PEG2-CH2CH2COOH is a Boc-protected, PEG-based linker for assembling PROTAC molecules. It combines a two-unit ethylene glycol spacer with a Boc-protected amino terminus and a terminal propionic acid, enabling versatile conjugation chemistry for connecting target ligands and E3 ligase ligands in small-molecule degraders.
  • Boc-protected amino terminus for selective deprotection and coupling.
  • Two-unit PEG spacer provides flexibility and appropriate linker length.
  • Terminal carboxylic acid enables standard amide or ester bond formation.
  • High purity suitable for research-grade synthesis workflows.
  • Good solubility in DMSO for easy stock solution preparation.