Learn More
Medchemexpress LLC Boc-NH-PEG2-CH2COOH | 108466-89-3 | 263.29 | 100 G
SDP
Catalog No. 5000445541
Encompass_Preferred
Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by a linker; one binds to an E3 ubiquitin ligase and the other to the target protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Enables selective degradation of target proteins
Catalog No. 50-004-45541 Supplier Medchemexpress LLC Supplier No. HYW005056100G
Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed with two different ligands connected by a linker; one binds to an E3 ubiquitin ligase and the other to the target protein, enabling the selective degradation of target proteins through the intracellular ubiquitin-proteasome system.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Enables selective degradation of target proteins