Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-NH-PEG2-CH2COOH | 108466-89-3 | 263.29 | 5 G
SDP

Catalog No. 5000445984
Encompass_Preferred

Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only and not sold to patients.

  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • For research use only
  • Molecular formula: C11H21NO6
  • Appearance: Liquid, colorless to light yellow
  • Storage: Pure form at -20°C for 3 years, 4°C for 2 years
  • Storage: In solvent at -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL

Catalog No. 50-004-45984 Supplier Medchemexpress LLC Supplier No. HYW0050565G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is for research use only and not sold to patients.

  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • For research use only
  • Molecular formula: C11H21NO6
  • Appearance: Liquid, colorless to light yellow
  • Storage: Pure form at -20°C for 3 years, 4°C for 2 years
  • Storage: In solvent at -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO at 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.