Learn More
Medchemexpress LLC Boc-NH-PEG2-CH2COOH | 108466-89-3 | 263.29 | 50 G
SDP
Catalog No. 5000448886
Encompass_Preferred
Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are composed of two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to the target protein. They leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize two ligands connected by a linker
  • One ligand binds to an E3 ubiquitin ligase
  • Other ligand binds to the target protein
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • High solubility in DMSO (100 mg/mL)
Catalog No. 50-004-48886 Supplier Medchemexpress LLC Supplier No. HYW00505650G
Boc-NH-PEG2-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are composed of two distinct ligands connected by a linker, one binding to an E3 ubiquitin ligase and the other to the target protein. They leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs utilize two ligands connected by a linker
  • One ligand binds to an E3 ubiquitin ligase
  • Other ligand binds to the target protein
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • High solubility in DMSO (100 mg/mL)