Learn More
Medchemexpress LLC Boc-NH-PEG2 (PROTAC Linker 11) | 139115-91-6 | 205.25 | 5 G
SDP
Catalog No. 5000449681
Encompass_Preferred
The product is a polyethylene glycol (PEG)-based linker used in the creation of PROTACs, which are molecules designed to selectively degrade target proteins. These PROTACs function by utilizing the intracellular ubiquitin-proteasome system.
  • It is a polyethylene glycol-based molecule.
  • It is used in the synthesis of protein degraders.
  • The purity is very high at 99.78%.
  • It is supplied as a colorless to light yellow liquid.
  • It comes with a data sheet, safety data sheet, and certificate of analysis.
  • It can be stored for long periods, up to three years in pure form at -20°c.
Catalog No. 50-004-49681 Supplier Medchemexpress LLC Supplier No. HYW0048965G
The product is a polyethylene glycol (PEG)-based linker used in the creation of PROTACs, which are molecules designed to selectively degrade target proteins. These PROTACs function by utilizing the intracellular ubiquitin-proteasome system.
  • It is a polyethylene glycol-based molecule.
  • It is used in the synthesis of protein degraders.
  • The purity is very high at 99.78%.
  • It is supplied as a colorless to light yellow liquid.
  • It comes with a data sheet, safety data sheet, and certificate of analysis.
  • It can be stored for long periods, up to three years in pure form at -20°c.