Learn More
Medchemexpress LLC Boc-NH-PEG3 | 139115-92-7 | MFCD07357496 | ≥98.0% | 249.30 | C11H23NO5 | 1 G
SDP
Catalog No. 5000301635
Encompass_Preferred
Boc-NH-PEG3 is a Boc-protected PEG3 (triethylene glycol) linker used as a short, hydrophilic spacer in the chemical synthesis of PROTACs and related bioconjugates. It provides a flexible, soluble spacer terminated with a Boc-protected amine that can be deprotected and further functionalized for coupling to ligands or tags. The compound is supplied at high purity and in research-scale pack sizes suitable for synthetic workflows.
  • Boc-protected terminal amine for orthogonal deprotection.
  • Triethylene glycol spacer gives flexibility and solubility.
  • Suitable for PROTAC and general bioconjugation chemistry.
  • High purity, suitable for synthetic applications.
  • Available in small pack sizes for research-scale synthesis.
  • Stable when stored under recommended conditions.
Catalog No. 50-003-01635 Supplier Medchemexpress LLC Supplier No. HYW0177721G
Add to Cart
Add to Cart
Boc-NH-PEG3 is a Boc-protected PEG3 (triethylene glycol) linker used as a short, hydrophilic spacer in the chemical synthesis of PROTACs and related bioconjugates. It provides a flexible, soluble spacer terminated with a Boc-protected amine that can be deprotected and further functionalized for coupling to ligands or tags. The compound is supplied at high purity and in research-scale pack sizes suitable for synthetic workflows.
  • Boc-protected terminal amine for orthogonal deprotection.
  • Triethylene glycol spacer gives flexibility and solubility.
  • Suitable for PROTAC and general bioconjugation chemistry.
  • High purity, suitable for synthetic applications.
  • Available in small pack sizes for research-scale synthesis.
  • Stable when stored under recommended conditions.