Learn More
Medchemexpress LLC Boc-NH-PEG3 | 139115-92-7 | MFCD07357496 | ≥98.0% | 249.30 | C11H23NO5 | 5 G
SDP
Catalog No. 5000301667
Encompass_Preferred
Boc-NH-PEG3 is a polyethylene glycol-based linker bearing a Boc-protected amino group and a triethylene glycol spacer. It is intended as a synthetic building block for assembling bifunctional degraders and other conjugates, providing aqueous solubility and a protected amine for downstream coupling reactions.
  • Boc-protected amine provides a protected functional handle for synthesis.
  • Triethylene glycol spacer increases solubility and conformational flexibility.
  • High purity suitable for chemical synthesis (≥98.0%).
  • Multiple package sizes available to support small- and large-scale work.
  • Storage recommendations extend shelf life when followed.
  • Liquid, colorless to light yellow, convenient for handling and formulation.
Catalog No. 50-003-01667 Supplier Medchemexpress LLC Supplier No. HYW0177725G
Boc-NH-PEG3 is a polyethylene glycol-based linker bearing a Boc-protected amino group and a triethylene glycol spacer. It is intended as a synthetic building block for assembling bifunctional degraders and other conjugates, providing aqueous solubility and a protected amine for downstream coupling reactions.
  • Boc-protected amine provides a protected functional handle for synthesis.
  • Triethylene glycol spacer increases solubility and conformational flexibility.
  • High purity suitable for chemical synthesis (≥98.0%).
  • Multiple package sizes available to support small- and large-scale work.
  • Storage recommendations extend shelf life when followed.
  • Liquid, colorless to light yellow, convenient for handling and formulation.