Learn More
Medchemexpress LLC Boc-nh-peg3 | 139115-92-7 | MFCD07357496 | 98.0% | 249.30 g/mol | C11H23NO5 | 10 G
SDP
Catalog No. 5000301666
Encompass_Preferred
Boc-NH-PEG3 is a Boc-protected PEG3-based linker used for assembling PROTACs and other bioconjugates. It provides a Boc-protected terminal amine and a terminal hydroxyl, offering orthogonal functional handles for stepwise coupling. The short PEG3 spacer improves aqueous solubility and imparts flexible spacing between linked ligands.
  • Boc-protected terminal amine enables selective deprotection and subsequent coupling.
  • Terminal hydroxyl allows activation or conjugation chemistry.
  • Short PEG3 spacer provides aqueous solubility and flexible linker geometry.
  • High stated purity (98.0%) supports reproducible synthesis.
  • Multiple pack sizes available for small-scale and preparative work.
Catalog No. 50-003-01666 Supplier Medchemexpress LLC Supplier No. HYW01777210G
Boc-NH-PEG3 is a Boc-protected PEG3-based linker used for assembling PROTACs and other bioconjugates. It provides a Boc-protected terminal amine and a terminal hydroxyl, offering orthogonal functional handles for stepwise coupling. The short PEG3 spacer improves aqueous solubility and imparts flexible spacing between linked ligands.
  • Boc-protected terminal amine enables selective deprotection and subsequent coupling.
  • Terminal hydroxyl allows activation or conjugation chemistry.
  • Short PEG3 spacer provides aqueous solubility and flexible linker geometry.
  • High stated purity (98.0%) supports reproducible synthesis.
  • Multiple pack sizes available for small-scale and preparative work.