Learn More
Cayman Chemical Boc-NH-PEG3-CH2CoH 100mg
SDP
Catalog No. 502990394
Encompass_Preferred
A building block; has been used as a linker in the synthesis of PROTACs targeting MEK; has been used as a spacer in the synthesis of radiolabeled probes for dual labeling of tumor integrin αVβ3 and GRPR in tumor tissue in vivo
Catalog No. 50-299-0394 Supplier Cayman Chemical Supplier No. 30536100 MG
A building block; has been used as a linker in the synthesis of PROTACs targeting MEK; has been used as a spacer in the synthesis of radiolabeled probes for dual labeling of tumor integrin αVβ3 and GRPR in tumor tissue in vivo