Learn More
Cayman Chemical Boc-NH-PEG3-OH 50mg
SDP
Catalog No. 502990187
Encompass_Preferred
A PEGylated building block; has been used as a spacer in biotinylated heterobifunctional cross-linker agents; has been used as a spacer in near infrared imaging probes for CYP1B1, which is expressed in a variety of tumors; has also been used in the synthesis of graftable peptidomimetics that bind to human αVβ3 integrin and inhibit adhesion of human osteoprogenitor cells to vitronectin
Catalog No. 50-299-0187 Supplier Cayman Chemical Supplier No. 3049650 MG
A PEGylated building block; has been used as a spacer in biotinylated heterobifunctional cross-linker agents; has been used as a spacer in near infrared imaging probes for CYP1B1, which is expressed in a variety of tumors; has also been used in the synthesis of graftable peptidomimetics that bind to human αVβ3 integrin and inhibit adhesion of human osteoprogenitor cells to vitronectin