Learn More
Medchemexpress LLC Boc-NH-PEG4 | 106984-09-2 | MFCD07779195 | 99.5% | 293.36 | C13H27NO6 | 5 G
SDP
Catalog No. 5000299702
Encompass_Preferred
Boc-NH-PEG4 is a Boc-protected polyethylene glycol (PEG4) linker used as a spacer in PROTAC, ADC, and other bioconjugation syntheses. It provides a four-unit PEG spacer with a Boc-protected amino terminus that enables orthogonal deprotection and downstream functionalization. The material is supplied as a colorless to light yellow liquid and is suitable as a building block in small-molecule conjugates.
  • Four-unit PEG spacer for controlled linker length.
  • Boc-protected amino terminus enables orthogonal deprotection and diversification.
  • High purity suitable for synthetic applications (about 99.5%).
  • Liquid form simplifies handling and dosing.
  • Recommended storage at -20°C to preserve stability.
  • Compatible with common coupling chemistries for conjugation workflows.
Catalog No. 50-002-99702 Supplier Medchemexpress LLC Supplier No. HYW0258965G
Boc-NH-PEG4 is a Boc-protected polyethylene glycol (PEG4) linker used as a spacer in PROTAC, ADC, and other bioconjugation syntheses. It provides a four-unit PEG spacer with a Boc-protected amino terminus that enables orthogonal deprotection and downstream functionalization. The material is supplied as a colorless to light yellow liquid and is suitable as a building block in small-molecule conjugates.
  • Four-unit PEG spacer for controlled linker length.
  • Boc-protected amino terminus enables orthogonal deprotection and diversification.
  • High purity suitable for synthetic applications (about 99.5%).
  • Liquid form simplifies handling and dosing.
  • Recommended storage at -20°C to preserve stability.
  • Compatible with common coupling chemistries for conjugation workflows.