Learn More
Medchemexpress LLC Boc-nh-peg4 | 106984-09-2 | MFCD07779195 | 99.5% | 293.36 g/mol | C13H27NO6 | 1 G
SDP
Catalog No. 5000301612
Encompass_Preferred
Boc-NH-PEG4 is a polyethylene glycol (PEG)-based linker with a Boc-protected amino group and a terminal hydroxyl designed for use in the synthesis of bifunctional molecules such as PROTACs. It provides a hydrophilic, flexible spacer that supports conjugation between two ligands while maintaining solubility and synthetic handle availability.
  • Provides a flexible PEG4 spacer for linking ligands.
  • Boc-protected amino group enables selective deprotection and functionalization.
  • Terminal hydroxyl offers options for further derivatization.
  • Hydrophilic character improves solubility in aqueous-organic media.
  • High purity supports consistent synthetic results.
Catalog No. 50-003-01612 Supplier Medchemexpress LLC Supplier No. HYW0258961G
Boc-NH-PEG4 is a polyethylene glycol (PEG)-based linker with a Boc-protected amino group and a terminal hydroxyl designed for use in the synthesis of bifunctional molecules such as PROTACs. It provides a hydrophilic, flexible spacer that supports conjugation between two ligands while maintaining solubility and synthetic handle availability.
  • Provides a flexible PEG4 spacer for linking ligands.
  • Boc-protected amino group enables selective deprotection and functionalization.
  • Terminal hydroxyl offers options for further derivatization.
  • Hydrophilic character improves solubility in aqueous-organic media.
  • High purity supports consistent synthetic results.