Learn More
Medchemexpress LLC Boc-NH-PEG4-C2-Boc | 2138484-13-4 | 421.53 | 100 MG
SDP
Catalog No. 5000349467
Encompass_Preferred
Boc-NH-PEG4-C2-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only
Catalog No. 50-003-49467 Supplier Medchemexpress LLC Supplier No. HY138370100MG
Boc-NH-PEG4-C2-Boc is a PEG-based PROTAC linker designed for the synthesis of PROTACs. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting a ligand for an E3 ubiquitin ligase with a ligand for the target protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • For research use only