Learn More
Medchemexpress LLC Boc-NH-PEG4-C2-Boc | 2138484-13-4 | 421.53 | 25 MG
SDP
Catalog No. 5000348643
Encompass_Preferred
Boc-NH-PEG4-C2-Boc is a PEG-based PROTAC linker used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). These molecules are designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system. PROTACs feature two different ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two ligands connected by a linker
  • Targets an E3 ubiquitin ligase
  • Targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-48643 Supplier Medchemexpress LLC Supplier No. 25MGHY138370
Boc-NH-PEG4-C2-Boc is a PEG-based PROTAC linker used in the synthesis of Proteolysis Targeting Chimeras (PROTACs). These molecules are designed to selectively degrade target proteins by exploiting the intracellular ubiquitin-proteasome system. PROTACs feature two different ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two ligands connected by a linker
  • Targets an E3 ubiquitin ligase
  • Targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system for selective protein degradation