Learn More
Medchemexpress LLC Boc-NH-PEG4-C2-Boc | 2138484-13-4 | 421.53 | 250 MG
SDP
Catalog No. 5000351563
Encompass_Preferred
Boc-NH-PEG4-C2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are compounds containing two different ligands connected by a linker: one ligand for an E3 ubiquitin ligase and the other for the target protein. They utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-51563 Supplier Medchemexpress LLC Supplier No. 250MGHY138370
Boc-NH-PEG4-C2-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs are compounds containing two different ligands connected by a linker: one ligand for an E3 ubiquitin ligase and the other for the target protein. They utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins