Learn More
Medchemexpress LLC HY-W008352 100mg Medchemexpress, Boc-NH-PEG4-CH2CH2NH2 CAS:811442-84-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW008352100MG
Encompass_Preferred
Medchemexpress, HY-W008352 100mg Boc-NH-PEG4-CH2CH2NH2 CAS:811442-84-9 Boc-NH-PEG4-CH2CH2NH2 a cleavable 5 unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3253