Learn More
Medchemexpress LLC Boc-NH-PEG4-CH2COOH | 876345-13-0 | MFCD30528568 | >97.0% | 351.39 g/mol | C15H29NO8 | 250 MG
SDP
Catalog No. 5000202882
Encompass_Preferred
Boc-NH-PEG4-CH2COOH is a Boc-protected PEG4 linker with a terminal carboxylic acid offered for research use. It is used as a cleavable linker in antibody-drug conjugates (ADCs) and as a PEG-based linker in PROTAC synthesis. The product is supplied in multiple package sizes and is provided with analytical purity appropriate for research applications.
  • Boc-protected PEG4 linker with terminal carboxylic acid.
  • Suitable for ADC and PROTAC linker applications.
  • High reported purity for research use.
  • Available in small-scale and bulk package sizes.
  • Molecular weight 351.39 g/mol; formula C15H29NO8.
Catalog No. 50-002-02882 Supplier Medchemexpress LLC Supplier No. HY42640250MG
Boc-NH-PEG4-CH2COOH is a Boc-protected PEG4 linker with a terminal carboxylic acid offered for research use. It is used as a cleavable linker in antibody-drug conjugates (ADCs) and as a PEG-based linker in PROTAC synthesis. The product is supplied in multiple package sizes and is provided with analytical purity appropriate for research applications.
  • Boc-protected PEG4 linker with terminal carboxylic acid.
  • Suitable for ADC and PROTAC linker applications.
  • High reported purity for research use.
  • Available in small-scale and bulk package sizes.
  • Molecular weight 351.39 g/mol; formula C15H29NO8.