Learn More
Medchemexpress LLC Boc-NH-PEG4-Ms | 1447797-72-9 | 98.0% | C14H29NO8S | 100 MG
SDP
Catalog No. 5000352939
Encompass_Preferred
Boc-NH-PEG4-MS is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These compounds utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is intended for laboratory research and the manufacture of substances.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Leverages the ubiquitin-proteasome system to degrade target proteins
  • Appears as a colorless to light yellow liquid
  • NMR and LCMS spectra consistent with structure
  • Stable in pure form: -20°C for 3 years; 4°C for 2 years
  • Stable in solvent: -80°C for 6 months; -20°C for 1 month
Catalog No. 50-003-52939 Supplier Medchemexpress LLC Supplier No. HY140391100MG
Boc-NH-PEG4-MS is a PEG-based PROTAC linker designed for the synthesis of PROTACs. These compounds utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. It is intended for laboratory research and the manufacture of substances.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Leverages the ubiquitin-proteasome system to degrade target proteins
  • Appears as a colorless to light yellow liquid
  • NMR and LCMS spectra consistent with structure
  • Stable in pure form: -20°C for 3 years; 4°C for 2 years
  • Stable in solvent: -80°C for 6 months; -20°C for 1 month