Learn More
Medchemexpress LLC Boc-NH-PEG4-Ms | 1447797-72-9 | 98.0% | C14H29NO8S | 250 MG
SDP
Catalog No. 5000352754
Encompass_Preferred
Boc-NH-PEG4-MS is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52754 Supplier Medchemexpress LLC Supplier No. 250MGHY140391
Boc-NH-PEG4-MS is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. These PROTACs degrade target proteins by exploiting the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand targets an E3 ubiquitin ligase
  • The other ligand targets a specific protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins