Learn More
Medchemexpress LLC Boc-NH-PEG5-CH2CH2COOH | 1347750-78-0 | MFCD24539478 | ≥97.0% | 409.47 g/mol | C18H35NO9 | 250 MG
SDP
Catalog No. 5000302740
Encompass_Preferred
Boc-NH-PEG5-CH2CH2COOH is a Boc-protected PEG5 linker terminating in a propionic acid functionality. It is used as a modular building block for assembling bifunctional molecules such as PROTACs, providing a hydrophilic, flexible spacer and a protected amine handle for selective deprotection and subsequent coupling reactions.
  • Hydrophilic PEG5 spacer improves solubility and flexibility.
  • Boc-protected amine allows controlled deprotection and functionalization.
  • Terminal carboxylic acid enables straightforward coupling to ligands or linkers.
  • Moderate molecular weight suitable for linker design without excessive bulk.
  • Intended for use as a chemical building block in medicinal chemistry workflows.
Catalog No. 50-003-02740 Supplier Medchemexpress LLC Supplier No. 250MGHYW022232
Boc-NH-PEG5-CH2CH2COOH is a Boc-protected PEG5 linker terminating in a propionic acid functionality. It is used as a modular building block for assembling bifunctional molecules such as PROTACs, providing a hydrophilic, flexible spacer and a protected amine handle for selective deprotection and subsequent coupling reactions.
  • Hydrophilic PEG5 spacer improves solubility and flexibility.
  • Boc-protected amine allows controlled deprotection and functionalization.
  • Terminal carboxylic acid enables straightforward coupling to ligands or linkers.
  • Moderate molecular weight suitable for linker design without excessive bulk.
  • Intended for use as a chemical building block in medicinal chemistry workflows.