Learn More
Medchemexpress LLC Boc-NH-PEG5-CH2CH2COOH | 1347750-78-0 | MFCD24539478 | 99.9% | 409.47 g/mol | C18H35NO9 | 100 MG
SDP
Catalog No. 5000301729
Encompass_Preferred
Boc-NH-PEG5-CH2CH2COOH is a Boc-protected PEG5 linker containing a protected amine and a terminal ethyl carboxylic acid, intended for use in the synthesis of PROTAC molecules and other linker-dependent conjugates. It is a mid-sized PEG derivative offering predictable reactivity for coupling chemistries and convenient handling in common organic solvents.
  • Boc-protected amine facilitates orthogonal deprotection
  • PEG5 spacer provides flexible linker length
  • Terminal ethyl carboxylic acid enables amide coupling
  • High solubility in DMSO for stock solutions
  • Stable under recommended cold storage conditions
Catalog No. 50-003-01729 Supplier Medchemexpress LLC Supplier No. HYW022232100MG
Boc-NH-PEG5-CH2CH2COOH is a Boc-protected PEG5 linker containing a protected amine and a terminal ethyl carboxylic acid, intended for use in the synthesis of PROTAC molecules and other linker-dependent conjugates. It is a mid-sized PEG derivative offering predictable reactivity for coupling chemistries and convenient handling in common organic solvents.
  • Boc-protected amine facilitates orthogonal deprotection
  • PEG5 spacer provides flexible linker length
  • Terminal ethyl carboxylic acid enables amide coupling
  • High solubility in DMSO for stock solutions
  • Stable under recommended cold storage conditions