Learn More
Medchemexpress LLC Boc-NH-PEG6-amine | 1091627-77-8 | 97.0% | C19H40N2O8 | 250 MG
SDP
Catalog No. 5000352753
Encompass_Preferred
Boc-NH-PEG6-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Utilized in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52753 Supplier Medchemexpress LLC Supplier No. 250MGHY140231
Boc-NH-PEG6-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs are molecules that leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • Utilized in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins