Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-NH-PEG6-CH2CH2COOH | 882847-13-4 | 97.0% | 453.52 | C20H39NO10 | 5 G
SDP

Catalog No. 5000203065
Encompass_Preferred

Boc-NH-PEG6-CH2CH2COOH is a Boc-protected PEG6-based cleavable linker used for antibody-drug conjugate and PROTAC linker synthesis. It provides a Boc-protected amine at one end and a terminal carboxylic acid at the other, enabling common bioconjugation strategies and linker design in medicinal chemistry.

  • Cleavable linker for ADC and PROTAC applications.
  • Boc-protected amine and carboxylic acid functional groups.
  • Molecular weight 453.52.
  • Chemical formula C20H39NO10.
  • Purity 97.0%.
  • Supplied as a viscous liquid in multiple package sizes, including 5 G.

Catalog No. 50-002-03065 Supplier Medchemexpress LLC Supplier No. HYW0402445G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-NH-PEG6-CH2CH2COOH is a Boc-protected PEG6-based cleavable linker used for antibody-drug conjugate and PROTAC linker synthesis. It provides a Boc-protected amine at one end and a terminal carboxylic acid at the other, enabling common bioconjugation strategies and linker design in medicinal chemistry.

  • Cleavable linker for ADC and PROTAC applications.
  • Boc-protected amine and carboxylic acid functional groups.
  • Molecular weight 453.52.
  • Chemical formula C20H39NO10.
  • Purity 97.0%.
  • Supplied as a viscous liquid in multiple package sizes, including 5 G.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.