Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-NH-PEG6-CH2COOH | 391684-36-9 | ≥98.0% | C19H37NO10 | 100 MG
SDP

Catalog No. 5000356197
Encompass_Preferred

Boc-NH-PEG6-CH2COOH is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. This compound is for research use only and not intended for patient use. It helps exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Molecular weight: 439.50
  • Appearance: Colorless to light yellow liquid
  • Density: 1.134±0.06 g/cm3
  • Shipping at room temperature (continental US)
  • Storage (pure form): -20°C for 3 years or 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-56197 Supplier Medchemexpress LLC Supplier No. HY140477100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-NH-PEG6-CH2COOH is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. This compound is for research use only and not intended for patient use. It helps exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Molecular weight: 439.50
  • Appearance: Colorless to light yellow liquid
  • Density: 1.134±0.06 g/cm3
  • Shipping at room temperature (continental US)
  • Storage (pure form): -20°C for 3 years or 4°C for 2 years
  • Storage (in solvent): -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.