Learn More
Medchemexpress LLC Boc-NH-PEG7-acetic acid | 141282-29-3 | MFCD08274604 | 98.0% | 483.55 g/mol | C21H41NO11 | 50 MG
SDP
Catalog No. 5000280730
Encompass_Preferred
Boc-NH-PEG7-acetic acid is a Boc-protected, PEG7-based linker bearing a terminal acetic acid used in medicinal chemistry for assembling proteolysis-targeting chimeras (PROTACs) and other bifunctional conjugates. It offers a flexible, hydrophilic spacer and a protected amine for stepwise synthetic transformations.
  • PEG7 spacer provides flexibility and improved solubility in conjugates.
  • Boc-protected amine allows selective deprotection and subsequent coupling.
  • Terminal acetic acid enables straightforward amide bond formation.
  • Reported high purity (98.0%) suitable for research synthesis.
  • Offered in multiple lab-scale pack sizes to support synthesis needs.
Catalog No. 50-002-80730 Supplier Medchemexpress LLC Supplier No. HYW19096350MG
Boc-NH-PEG7-acetic acid is a Boc-protected, PEG7-based linker bearing a terminal acetic acid used in medicinal chemistry for assembling proteolysis-targeting chimeras (PROTACs) and other bifunctional conjugates. It offers a flexible, hydrophilic spacer and a protected amine for stepwise synthetic transformations.
  • PEG7 spacer provides flexibility and improved solubility in conjugates.
  • Boc-protected amine allows selective deprotection and subsequent coupling.
  • Terminal acetic acid enables straightforward amide bond formation.
  • Reported high purity (98.0%) suitable for research synthesis.
  • Offered in multiple lab-scale pack sizes to support synthesis needs.