Learn More
Medchemexpress LLC Boc-NH-PEG7-acid | 2055044-68-1 | C22H43NO11 | 250 MG
SDP
Catalog No. 5000352742
Encompass_Preferred
Boc-NH-PEG7-acid is a PEG-based PROTAC linker primarily utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52742 Supplier Medchemexpress LLC Supplier No. 250MGHY140469
Boc-NH-PEG7-acid is a PEG-based PROTAC linker primarily utilized in the synthesis of PROTACs. PROTACs are designed with two distinct ligands connected by a linker: one targets an E3 ubiquitin ligase, and the other targets a specific protein. This mechanism allows PROTACs to leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the ubiquitin-proteasome system
  • Selectively degrades target proteins