Learn More
Medchemexpress LLC Boc-NH-PEG7-propargyl | 2112737-90-1 | >=97.0% | C22H41NO9 | 100 MG
SDP
Catalog No. 5000355026
Encompass_Preferred
Boc-NH-PEG7-propargyl is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-55026 Supplier Medchemexpress LLC Supplier No. HY140882100MG
Boc-NH-PEG7-propargyl is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an alkyne group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups