Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-NH-PEG8-C2-Br | 2688072-12-8 | 99.8% | 576.52 | 25 MG
SDP

Catalog No. 5000352441
Encompass_Preferred

Boc-NH-PEG8-C2-Br is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is intended for research use only.

  • Molecular weight: 576.52
  • Chemical formula: C23H46BrNO10
  • Appearance: Colorless to light yellow liquid
  • Purity (HPLC): 99.8%
  • Consistent with structure by 1H NMR and MS
  • Applicable in cancer and cancer targeted therapy research

Catalog No. 50-003-52441 Supplier Medchemexpress LLC Supplier No. 25MGHY138406
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-NH-PEG8-C2-Br is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is intended for research use only.

  • Molecular weight: 576.52
  • Chemical formula: C23H46BrNO10
  • Appearance: Colorless to light yellow liquid
  • Purity (HPLC): 99.8%
  • Consistent with structure by 1H NMR and MS
  • Applicable in cancer and cancer targeted therapy research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.