Learn More
Medchemexpress LLC Boc-NH-PEG8-C2-Br | 2688072-12-8 | 99.8% | 576.52 | 50 MG
SDP
Catalog No. 5000349348
Encompass_Preferred
Boc-NH-PEG8-C2-Br is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins. This product is for research use only and not sold to patients.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Targets protein degradation via ubiquitin-proteasome system
  • For research use only
Catalog No. 50-003-49348 Supplier Medchemexpress LLC Supplier No. HY13840650MG
Boc-NH-PEG8-C2-Br is a PEG-based PROTAC linker. It is used in the synthesis of PROTACs, which are molecules designed to leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins. This product is for research use only and not sold to patients.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Targets protein degradation via ubiquitin-proteasome system
  • For research use only