Learn More
Medchemexpress LLC Boc-NH-Tri-(carbonylethoxymethyl)-methane | 220431-61-8 | MFCD34178797 | 98.4% | 437.44 g/mol | C18H31NO11 | 25 MG
SDP
Catalog No. 5000282219
Encompass_Preferred
Boc-NH-Tri-(carbonylethoxymethyl)-methane is a Boc-protected branched PEG linker bearing a protected amino group and three terminal carboxylic acid functionalities. It is intended for use as a building block in bioconjugation and linker design; the Boc group can be removed under mild acidic conditions to reveal a free amine for subsequent coupling reactions. Supplied at laboratory scale for synthetic applications.
  • Boc-protected amino group for controlled deprotection.
  • Three terminal carboxylic acids for multi-point conjugation.
  • Branched PEG spacer increases hydrophilicity and solubility.
  • Suitable for bioconjugation and linker synthesis.
  • High purity suitable for synthetic work (98.4%).
  • Molecular weight 437.44 g/mol; formula C18H31NO11.
Catalog No. 50-002-82219 Supplier Medchemexpress LLC Supplier No. HYW19095825MG
Boc-NH-Tri-(carbonylethoxymethyl)-methane is a Boc-protected branched PEG linker bearing a protected amino group and three terminal carboxylic acid functionalities. It is intended for use as a building block in bioconjugation and linker design; the Boc group can be removed under mild acidic conditions to reveal a free amine for subsequent coupling reactions. Supplied at laboratory scale for synthetic applications.
  • Boc-protected amino group for controlled deprotection.
  • Three terminal carboxylic acids for multi-point conjugation.
  • Branched PEG spacer increases hydrophilicity and solubility.
  • Suitable for bioconjugation and linker synthesis.
  • High purity suitable for synthetic work (98.4%).
  • Molecular weight 437.44 g/mol; formula C18H31NO11.