Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Boc-Nva-OH | 53308-95-5 | 98.0% | 217.26 | 25 G
SDP

Catalog No. 5000366933
Encompass_Preferred

Boc-Nva-OH is a valine derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress related tasks and prevent exercise induced muscle damage. They are recognized to be beneficial as ergogenic dietary substances.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping at room temperature in continental US; may vary elsewhere.
  • Storage as powder: -20°C for 3 years, 4°C for 2 years.
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month.
  • Related to peptides.
  • Reagent for peptide synthesis.
  • Related to amino acids and derivatives.
  • Available documentation: data sheet, SDS, COA, HNMR, and handling instructions.

Catalog No. 50-003-66933 Supplier Medchemexpress LLC Supplier No. 25GHYW012446
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Boc-Nva-OH is a valine derivative. Amino acids and amino acid derivatives have been commercially used as ergogenic supplements. They influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress related tasks and prevent exercise induced muscle damage. They are recognized to be beneficial as ergogenic dietary substances.

  • Appearance: Solid
  • Color: White to off-white
  • Shipping at room temperature in continental US; may vary elsewhere.
  • Storage as powder: -20°C for 3 years, 4°C for 2 years.
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month.
  • Related to peptides.
  • Reagent for peptide synthesis.
  • Related to amino acids and derivatives.
  • Available documentation: data sheet, SDS, COA, HNMR, and handling instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.