Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boc-ON, 98+%

Catalog No. AAA1890622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1890622 100 g
AAA1890606 5 g
AAA1890614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1890622 Supplier Thermo Scientific Chemicals Supplier No. A1890622
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 58632-95-4 | C13H14N2O3 | 246.266 g/mol

Widely used for protection of amino groups in peptide synthesis and multi-step organic synthesis of small molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Widely used for protection of amino groups in peptide synthesis and multi-step organic synthesis of small molecules.

Solubility
Soluble in ethyl acetate, ether and benzene. Insoluble in water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58632-95-4
Molecular Formula C13H14N2O3
Molecular Weight (g/mol) 246.266
MDL Number MFCD00001863
InChI Key QQWYQAQQADNEIC-PTNGSMBKSA-N
Synonym 2-tert-butoxycarbonyloxyimino-2-phenylacetonitrile, 2-boc-oxyimino-2-phenylacetonitrile, boc-on, ccris 2601, tert-butyl alpha-cyanobenzylaminyl carbonate, 2-tert-butoxycarbonyloximino-2-phenylacetonitrile, carbonic acid, cyanophenylmethylene azanyl 1,1-dimethylethyl ester, n-tert-butoxycarbonyl oxy benzimidoyl cyanide, benzeneacetonitrile, alpha-1,1-dimethylethoxy carbonyl oxy imino
PubChem CID 5868400
IUPAC Name tert-butyl [(E)-[cyano(phenyl)methylidene]amino] carbonate
SMILES CC(C)(C)OC(=O)ON=C(C#N)C1=CC=CC=C1

Specifications

Melting Point 86°C to 89°C
Quantity 100 g
UN Number UN3439
Beilstein 2217296
Sensitivity Moisture sensitive
Solubility Information Soluble in ethyl acetate,ether and benzene. Insoluble in water.
Formula Weight 246.27
Percent Purity ≥98%
Chemical Name or Material Boc-ON
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 261-370-9
RTECSNumber DA0513600
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.