Learn More
Cayman Chemical Boc-Orn(Fmoc)-OH, Purity: ≥98%, CAS: 150828-96-9, 500 mg
SDP
Catalog No. 502991347
Encompass_Preferred
An amino acid-containing building block; has been used to introduce δ-linked ornithine turns into synthetic macrocyclic β-sheets
Catalog No. 50-299-1347 Supplier Cayman Chemical Supplier No. 30495500 MG
An amino acid-containing building block; has been used to introduce δ-linked ornithine turns into synthetic macrocyclic β-sheets