Learn More
Medchemexpress LLC Boc-PEG4-C2-NHS ester | 1835759-75-5 | C20H33NO10 | 250 MG
SDP
Catalog No. 5000352591
Encompass_Preferred
Boc-PEG4-C2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Utilizes two ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52591 Supplier Medchemexpress LLC Supplier No. 250MGHY141122
Boc-PEG4-C2-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Utilizes two ligands connected by a linker
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins